Graded Seals, Quartz to Borosilicate, Ace Glass Incorporated
Fournisseur: Ace Glass
Certificats
A propos de cet article
For connecting Kimax, Pyrex, Simax or Duran 33 Expansion borosilicate glass parts to quartz
Quartz length is 7.6 cm, glass length, 10.2 cm.
Customers Who Bought This Also Bought