Stirring Bearing, Vacuum, Ace Glass Incorporated
Fournisseur: Ace Glass
Certificats
A propos de cet article
The upper member is internally threaded on the top
- Easier to adjust shaft
- suitable for vacuum work
- For use with ACE stirring shafts
#7 Ace‑Thred nylon bushing and FETFE o-ring form a compression seal on stirring shaft in place of the conventional tubing arrangement.
Customers Who Bought This Also Bought