Palladium standard solution, 10000 mg/L Pd in 10% hydrochloric acid (from Pd) ARISTAR® standard for ICP
# de catalogue 89418-734
Fournisseur: Prosci
Numéro CAS:
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
- Species:Human
- Size:0.05 mg
- Storage conditions:Store at –20 °C, stable for one year.
- Amino acid number:13 aa in the center
- Protein/peptide name:PAK6 Peptide (13 aa in the center)
- Tested applications:Blocking
- Cat. no.:89418-734
- Supplier No.:3073P
Spécifications
A propos de cet article
PAK6 peptide is used for blocking activity of PAK6 antibody.
Customers Who Bought This Also Bought