SP Bel-Art H-B DURAC® Plus™ Pocket Liquid-In-Glass Thermometers, Organic Liquid Fill, Bel-Art Products, a part of SP
Fournisseur: Bel-Art Products, a Part of SP
Certificats
A propos de cet article
Featuring a case to protect from breakage, these total immersion thermometers are a compact 160mm (6.3”), making them ideal for field studies, waste water treatment and any “on-the-go” applications
Each total immersion thermometer features a 160mm (6.3”) length and comes with a Statement of Accuracy indicating accuracy traceable to NIST. They are individually serialized and packaged in recyclable Safety-Pak™ packaging to prevent breakage.
- Mercury free; contain blue organic liquid against lead-free, white back glass
- Tested and inspected by our USA quality team
- Traceable to NIST
Case options include Plastic Window - encased in plastic with an open window, Closed Plastic - housed in removable plastic case, and Closed Metal - housed in a removable metal case.
Ordering information: Includes multi-language (English, French, German, Italian, Portuguese, and Spanish) Statement of Accuracy and Traceability.
Informations connexes
Services: VWRCATALYST™ offers convenient and efficient re-calibration services for your liquid in glass thermometers with an industry leading 5-day average turn-around time from receipt of your instrument. Simply order the stated catalog no. based on how many calibration points are desired. You will receive a protective box to ship your instrument to our certified calibration laboratory along with a work order form to specify your calibration points 0 to 100°C (32 to 212°F) and prepaid return shipping label. Upon receipt at the laboratory your instrument will be inspected for any defect under a microscope and the performance of your instrument will be compared to certified standards and corrections to one-tenth of the smallest division recorded on your signed calibration report. Your calibration report and instrument are carefully repacked then returned to you prepaid.