SP Bel-Art Enviro-Safe® Pocket Liquid-in-Glass Thermometers with Biodegradable Liquid, Bel-Art Products, a part of SP
Fournisseur: Bel-Art Products, a Part of SP
Certificats
A propos de cet article
EnviroKleen™ certified thermometers feature green, non-toxic, biodegradable, Enviro-Safe® liquid against lead-free, white back glass
Each thermometer features a 160mm (6.3') length, is total immersion and comes with a Statement of Accuracy indicating accuracy traceable to NIST and DKD. They are packaged in recyclable Safety-Pak™ packaging to prevent breakage.
- Total immersion pocket thermometers are a compact 160mm (6.3”) long
- Come with a case to protect from breakage
- Ideal for field studies, waste water treatment and any “on-the-go” applications
- Manufactured and tested by our USA quality team
- Traceable to NIST
Should you require an individualized Thermometer Calibration Report, custom calibration services are available.
Certifications: Each item is tested and calibrated in our exclusive triple accredited/registered ISO/IEC 17025:2005, A2LA accredited laboratory against equipment whose calibration is traceable to NIST.
Ordering information: Include a multi-language (English, French, German, Italian, Portuguese, and Spanish) Statement of Accuracy and Traceability.
Spécifications
- Catalog No:
- 30617-822
- 30617-812
- 30617-834
- 30617-828
- Environmentally Preferable:
- Safer Human and Environmental Health
- Safer Human and Environmental Health
- Safer Human and Environmental Health
- Safer Human and Environmental Health
Spécifications
Informations connexes
Services: VWRCATALYST™ offers convenient and efficient re-calibration services for your liquid in glass thermometers with an industry leading 5-day average turn-around time from receipt of your instrument. Simply order the stated catalog no. based on how many calibration points are desired. You will receive a protective box to ship your instrument to our certified calibration laboratory along with a work order form to specify your calibration points 0 to 100°C (32 to 212°F) and prepaid return shipping label. Upon receipt at the laboratory your instrument will be inspected for any defect under a microscope and the performance of your instrument will be compared to certified standards and corrections to one-tenth of the smallest division recorded on your signed calibration report. Your calibration report and instrument are carefully repacked then returned to you prepaid.