Titrette® Bottle Top Burets, Chemglass
Fournisseur: Chemglass
Certificats
A propos de cet article
The CG-1161 series Titrette® is the new generation of bottle top burette from BRAND
Features include Class A precision, Smooth, Low-force operation, low-profile structure, and chemical compatibility.
It has a number of innovative features and upgrades for more accurate and convenient titration. The Titrette® avoids the risk of spills from poured transfers to glass or plastic burets. The instrument is well suited for general chemistry, water treatment applications, food and beverage analysis, industrial titrations and environmental work.
Ordering information: Complete with a bottle top buret with a GL-45 thread, telescoping filling tube 170-330 mm in length, recirculation tube, 2 batteries, 2 colored light shield inspection windows, three bottle adapters with GL45 to 33-400 GPI thread, GL45 to S40 European socket adapter and GL45 to 29/32 inner joint, operating manual and 1 year warranty.
Customers Who Bought This Also Bought