SP Bel-Art Hydrometer Jar, Bel-Art Products, a part of SP
Catalog # 47750-918
Supplier: Bel-Art Products, a Part of SP
Specifications
- Ø:3.3 cm ( 1⅝")
- Capacity:245 ml (8.3 oz.)
- Description:Hydrometer Jar
- Height:30.5 cm (12")
- Cat. no.:47750-918
- Supplier No.:178170000
Specifications
About this item
Hydrometer Jar is composed of two pieces - a clear, polycarbonate cylinder and a removable hexagonal polypropylene base.
- Clear polycarbonate cylinder
- Polypropylene base
- Broad base prevents tipping
Clarity of polycarbonate cylinder makes reading easy
Hydrometer not included.
Broad polyethylene base helps prevent tipping. The clear, unbreakable polycarbonate cylinder has a 245 mL capacity and the 4” polyethylene base is removable.
Customers Who Bought This Also Bought