SP Bel-Art Spray Pump Bottles, Bel-Art Products, a part of SP
Fournisseur: Bel-Art Products, a Part of SP
Certificats
A propos de cet article
Low-density plunger style polyethylene bottle produces a fine mist spray.
- LDPE construction
- Plunger-style spray pump design
- Clear cap
Includes clear cap to help prevent evaporation and accidental leaks.
Available in two sizes.
Customers Who Bought This Also Bought