Foil Rolls
Fournisseur: Heathrow Scientific
Certificats
A propos de cet article
Custom-sized pieces of foil can easily be created for covering small surfaces such as beakers, flasks, tubes, and petri dishes.
- Create custom-size pieces of foil
- Two sizes available
These foil rolls are ideal for autoclaving, freezing, incubating, protecting lab ware, lining, sealing, and general storage.
Customers Who Bought This Also Bought