Microscope slide Mailer for 5 Slides, End Opening
# de catalogue 82003-433
Fournisseur: Heathrow Scientific
Spécifications
- Color:Natural
- Description:End-opening 5 slide mailers
- Cat. no.:82003-433
- Supplier No.:HS15986V
Spécifications
A propos de cet article
Ideal for safe and secure transport of standard microscope slides, 76×26 mm (3×1")
- Polypropylene container holds five standard slides
- Inside wall features slotted channels to keep slides separated and to make removal easier
- Lid closes securely to keep slides safe
- Disposable
Dimension (L×W×H): 43×24×88 mm
Ordering information: Red, yellow, blue, purple, and orange Slide Mailers are available. Contact your Avantor sales representative for additional information.
Customers Who Bought This Also Bought