VWR® Tall Microscope Slide Boxes for 100 Slides
# de catalogue 89140-280
Fournisseur: VWR International
Spécifications
- Dimensions:208W x 175H x 60D mm (8⅛ x 6⅞ x 2⅜")
- Color:Blue
- Description:VWR® Tall Microscope Slide Boxes, 100-Place
- Cat. no.:89140-280
Spécifications
A propos de cet article
Tall slide boxes are designed for storage and transport of non-standard microscope slides (3x2").
- Grooved slots
- Soft cork lining
- Sturdy ABS construction for rugged use
Grooved slots hold individual slides in place. The box lid provides the needed clearance for the taller height of non-standard slides. Soft cork lining insures that your slides will not slip. Sturdy ABS construction for rugged use.
Customers Who Bought This Also Bought