Sodium hydrogen carbonate 99.7-100.3%, powder ACS
# de catalogue CA71008-496
Fournisseur: VWR
Numéro CAS: 144-55-8
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
- Packaging:Poly bottle
- Size:500 g
- Cat. no.:CA71008-496
- Supplier No.:80500-300
Spécifications
Résultats des tests de spécification
- Assay (dried basis) NaHCO3:99.7 - 100.3%
- Insoluble Matter:0.015% max.
- Chloride (Cl):0.003% max.
- Phosphate (PO4):0.001% max.
- Sulfur compounds (as SO4):0.003% max.
- Ammonium (NH4):5 ppm max.
- Heavy metals (as Pb):5 ppm max.
- Iron (Fe):0.001% max.
- Calcium (Ca):0.02% max.
- Magnesium (Mg):0.005% max.
- Potassium (K):0.005% max.
- Sodium hydrogen carbonate 99.7-100.3%, powder ACS:
Résultats des tests de spécification
A propos de cet article
Synonymes: Sodium bicarbonate
Point de Fusion:270 °C
Densité:2.159 g/cm³ (20 °C)
:Length:
:Width:
:Height:
:Ø
:Thickness:
:99,7-100,3
:Boiling Pt:
:Boiling Pt:
:Melting Pt:
:Melting Pt: 270 °C
:Flash Pt:
:Flash Pt:
:M.W. 84.01 g/mol
Index Merck:13,08655
Formule:NaHCO₃
MW:84.01 g/mol
Température d'entreposage:Ambient
Numéro MDL:MFCD00003528
Numéro CAS:144-55-8
EINECS:205-633-8
Index Merck:12,08726
Customers Who Bought This Also Bought