Platinum-Cobalt Colour Standard, Anachemia
Fournisseur: VWR
Certificats
A propos de cet article
These colour standards are classified as No
Visual Absorbance for a 1 cm cell vs H2O is:
0.110 - 0.120 at 430 nm
0.130 - 0.145 at 455 nm
0.105 - 0.120 at 480 nm
0.055 - 0.065 at 510 nm
500 Hazen.
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.