Nitric acid 68 - 70% ACS
Fournisseur: Avantor
Certificats
A propos de cet article
Synonymes: Nitric acid solution
Formule:HNO₃
MW:63.01 g/mol
Point d'ébullition:120.5 °C (1013 hPa)
Point de Fusion:-42 °C
Densité:1.4 g/cm³ (20 °C)
Numéro MDL:MFCD00011349
Numéro CAS:7697-37-2
Index Merck:13,06608
:63.01
ADR:8,II
:Length:
:Width:
:Height:
:Ø
:Thickness:
:Boiling Pt:
:Boiling Pt: 87 °C (1013 hPa)
:Melting Pt:
:Melting Pt: -42 °C
:Flash Pt:
:Flash Pt:
:Density: 1.51 g/cm³ (20 °C)
:M.W. 63.01 g/mol
:Nitric acid
Température d'entreposage:Ambient
EINECS:231-714-2
:-42
ONU:2031
:8
:II
:1.51
:GHS03
:GHS05
:12,06672
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Résultats des tests de spécification
- Appearance:Clear, colourless, free from suspended matter or sediment
- Assay (HNO3):68.0 -70.0%
- Color (APHA):10 ppm max.
- Residue after ignition:5 ppm max.
- Chloride (Cl):0.5 ppm max.
- Sulfate (SO4):1 ppm max.
- Arsenic (As):0.01 ppm max.
- Heavy metals (as Pb):0.2 ppm max.
- Iron (Fe):0.2 ppm max.
- Nitric acid 68 - 70% ACS:
Résultats des tests de spécification
Customers Who Bought This Also Bought