Shell Vials, Borosilicate Glass, with Plug-Style Needle Closure, Kimble Chase
Supplier: DWK Life Sciences (KIMBLE)
Certificates
About this item
TITESEAL® shell vials are ideal for chromatography applications.
- White polyethylene plug style needle closures, unattached
- Ideal for use with light-sensitive samples
- Made of 51 expansion borosilicate amber glass
Certifications: Meets USP Type I and ASTM E438, Type I, Class B requirements
Customers Who Bought This Also Bought