Cryo Marker Pen Set, Thermo Scientific
Catalog # 97049-678
Supplier: Thermo Fisher Scientific
Specifications
- Color:Black
- Description:Cryo pens, set of 4, all black
- Cat. no.:97049-678
- Supplier No.:4000222
Specifications
About this item
For fast-drying, non-smearing labeling
Recommended for all cryogenic vials at temperatures 200°C (-328°F)
Customers Who Bought This Also Bought