LabLink Power Strips, PG LifeLink
Fournisseur: PG Lifelink
Certificats
A propos de cet article
LabLink Power Strips are made of high quality extruded aluminum and can be permanently mounted to your work station.
- Extruded from #6063-T5 Aluminum Alloy
- Dimensions 1³/₄×2³/₄×18" - 60"
- Ideal for Mobile Benches
- Single Circuit Configuration
- Clear Satin Anodized Finish is Durable and Corrosion Resistant
- 20 Amp (NEMA 5-20R) Grey Duplex Receptacles
- 8 Foot, 3 Conductor, Black Power Cord with Straight Blade Plug (NEMA5-20P)
Power strips are available in lengths of 18", 24", 36", 48" and 60", providing more functionality for today's modular lab environment. Cover with 2 end plates.
Certifications: C-UL Listed.
Ordering information: Not to be used in conjunction with an extension cord or another power strip.
Customers Who Bought This Also Bought