LabLink Surface Mount Raceway, Versa-Duct, PG LifeLink
Fournisseur: PG Lifelink
Certificats
A propos de cet article
LabLink Surface Mount Raceway (Versa-Duct), made from anodized extruded aluminum, offers a superior design to combine electrical and/or data into one unified system.
- Extruded from #6063-T5 Aluminum Alloy
- Single Channel Assembly (1³/₄×2³/₄×48 – 120")
- Double Channel Assembly (2¹/₄×5¹/₄×48 – 120")
- Single Circuit Configuration
- 12" Pigtails are provided for Electrical Connections
- Clear Satin Anodized Finish is Durable and Corrosion Resistant
- 12" Removable Cover Plates for Easy Access in the Field
- 20 Amp (NEMA-5-20R) Grey Duplex Receptacles
- Grey Communication Holder with 2 Cat 5e Data Jacks (not wired)
- C-UL Listed
Factory pre-cut and pre-wired to standard lengths. Single channel raceway is configured to include power receptacles and dual channel raceway will include power and data. The raceway is provided in a clear satin anodized finish. Versa-Duct multi-outlet assemblies save you time and money by reducing on-site labor costs by up to 70%. Whether designing a raceway system for your laboratory work bench or a surface mounted system for an entire building, Versa-Duct is the raceway for you. Cover with two end plates.
Certifications: cUL Listed.
Customers Who Bought This Also Bought