Pyrite Crystal Group
# de catalogue 470210-868
Fournisseur: ADAM MOORE MO
Spécifications
- Description:Pyrite Crystal Group
- Cat. no.:470210-868
Spécifications
A propos de cet article
Choice intergrown crystal groups, many with striated pyritohedral crystal faces. Perfect for classroom study. Average size: 1" x 1". Peru.
Customers Who Bought This Also Bought