SP Bel-Art Sample Vials with Captive Closure, LDPE, Bel-Art Products, a part of SP
Fournisseur: Bel-Art Products, a Part of SP
Certificats
A propos de cet article
Cylindrical, low-density PE vials with friction-fit, captive closures open and close with a flick of your thumb.
- LDPE construction
- Captive closures controlled with thumb pressure
- Suited for storing liquids or storing and shipping dry materials
Excellent for storage of liquids as well as for storing and shipping dry materials.
Customers Who Bought This Also Bought