SP Bel-Art Measure-Matic™ Dispenser, Bel-Art Products, a part of SP
# de catalogue 66175-154
Fournisseur: Bel-Art Products, a Part of SP
Spécifications
- Volume:5 - 25 ml
- Description:Measure-Matic™ Automatic Measuring Liquid dispenser
- Supplier No.:116540000
Spécifications
A propos de cet article
Squeezing forces the desired volume of liquid through the tube and into the cup.
- Polyethylene storage bottle
- Polypropylene dispensing cup and delivery tube
- Hollow stopper serves as a dust cover
When pressure is released, excess fluid returns into the bottle for spill-free pouring.
Polypropylene dispensing cup slides onto a polypropylene delivery tube which is set in a low-density polyethylene 500 mL (16 oz.) storage bottle. A supplied hollow stopper serves as a dust cover.
Customers Who Bought This Also Bought