VWR® Traceable® Memory-Card Humidity/Temperature/Dew Point Meter | Avantor
VWR® Traceable® Memory-Card Humidity/Temperature/Dew Point Meter
Memory-Card Humidity/Temperature/Dew Point Meter, –0…50° C, ±1 °C, 0.0 to 90.0% RH
Catalog # 89369-158
Supplier:   VWR International
VWR® Traceable® Memory-Card Humidity/Temperature/Dew Point Meter
Catalog # 89369-158
Supplier:  VWR International
VWR® Traceable® Memory-Card Humidity/Temperature/Dew Point Meter
VWR® Traceable® Memory-Card Humidity/Temperature/Dew Point Meter
Memory-Card Humidity/Temperature/Dew Point Meter, –0…50° C, ±1 °C, 0.0 to 90.0% RH
Price
$1,069.75
Prix promotionnel pour les nouveaux clients
Log in pour la disponibilité
Each
Product Details & Documents
Traceable® Memory-Card Humidity/Temperature/Dew Point meter is a stand-alone unit with data logging capabilities.
  • Memory feature stores minimum/maximum readings
  • Visual and audio alarm features
  • Bright LED display
  • Unique 4-line display
It records readings from once a minute up to once every 720 minutes on a removable standard SD memory card
Minimum/maximum feature displays highest and lowest readings for any time period. Alarm feature sounds and flashes a bright red LED when temperature, humidity and/or dew points rise above or fall below user-set trip points. Alarm sets in 0.1 increments. Unique 4-line display simultaneously shows ambient temperature, humidity, dew point and time of day.
Memory card reader (supplied) plugs into any USB computer port for an instant report. Text report includes date/time of day and readings of sensors and can be sent to any spreadsheet or database. Unit monitors cabinets, incubators, clean rooms, store rooms, labs, hoods, and critical areas. Unit captures 5.9 million readings with the supplied 256 mb removable SD card.
Certifications: To assure accuracy, an individually serial-numbered Traceable® Certificate is provided from an ISO 17025 calibration laboratory accredited by A2LA. It indicates traceability to standards provided by NIST.
Ordering information: Supplied with probe, cable, 256 mb SD memory card, USB memory card reader, magnetic mounting strips, Velcro®, wall mount, bench stand, batteries, adaptor, Traceable® certificate.
Services: VWRCATALYST™ provides convenient and efficient re-calibration services for your digital hygrometers with an industry leading 5-day average turn-around time from receipt of your instrument. All the service will be performed by an ISO17025 certified lab. Recalibration, certification and shipping fees are all included in the catalog number cost. You will receive a protective box to ship your instrument to Avantor's certified calibration laboratory along with a work order form and a prepaid return shipping label. Upon receipt at the laboratory your instrument will be inspected for any defects and the performance of your instrument will be compared to certified standards. After re-calibration, your calibration report and instrument will be carefully repacked then returned with standard ground shipping. We also provide expedited service options available upon request. Hygrometer not included.
Spécifications
  • Plage de température:
    –0…50° C
  • Désignation:
    Memory-Card Humidity/Temperature/Dew Point Meter
  • Humidity Range:
    0.0 to 90.0% RH
  • Temperature Accuracy:
    ±1 °C
  • Humidity accuracy:
    ±5% RH at 25°C from 20 to 85% RH, ±7% elsewhere
  • Cat. No.:
    89369-158
  • Humidity resolution:
    0.1
  • Dew point range:
    –20.0…50.0 °C
  • Dew point resolution:
    0.1°
  • Temperature resolution:
    0.1°