SteriPettes, Kimble Chase, DWK Life Sciences
Supplier: DWK Life Sciences (KIMBLE)
Certificates
About this item
SteriPettes are sterile, disposable Pasteur pipettes produced from soda-lime, gamma-irradiated glass.
These pipettes are either non-plugged or cotton-plugged.
Customers Who Bought This Also Bought