Tygon® Laboratory Vacuum Tubing, Saint-Gobain
# de catalogue 89403-860
Fournisseur: Saint Gobain Life Sciences
Spécifications
- Length:50 ft.
- Color:Clear
- Description:Tygon® E-3603 Laboratory Vacuum Tubing
- Cat. no.:89403-860
- I.D.×O.D.:³⁄₁₆×⁵⁄₁₆"
- Supplier No.:ACF00012-V
Spécifications
A propos de cet article
The bio-based Tygon® line combines the high performance standards customers demand with an eco-friendly tubing design
Crystal clear and flexible, long-lasting and crack-resistant. Tygon tubing handles most inorganic chemicals found in the laboratory. It is non-oxidizing and non-contaminating, and less permeable than rubber tubing. The glassy-smooth inner bore helps prevent build-up to facilitate cleaning. Coils are marked at 1 foot or 30.4 cm intervals for easy measuring.
Customers Who Bought This Also Bought