ContactUsLinkComponent
Eisco® Electric Tuning Fork
Eisco® Electric Tuning Fork
# de catalogue 470138-868
Fournisseur:  Wards
TUNING FORK ELECTRIC
Eisco® Electric Tuning Fork
# de catalogue 470138-868
Fournisseur:  Wards
Spécifications
  • Description:
    Electric Tuning Fork
  • Cat. no.:
    470138-868
  • Supplier No.:
    PH0746-CNC
Spécifications
A propos de cet article
Show relationships between frequency, length, tension and mass
  • Heavy Duty
  • Repeatability
Learn about Melde's standing wave experiments with this fork. An electromagnet drives the fork to a sustained frequency of approx. 80Hz. One prong of the tuning fork has a captive knurled thumbscrew for performing Melde's experiment. Amplitude of the tuning fork vibrations is varied by means of a threaded adjusted screw. Universal binding posts are finished to provide electrical power from an external source of 2 to 6 volts DC, required but not included.
Customers Who Bought This Also Bought