VWR® Cuvette Holder, PE
# de catalogue 89000-626
Fournisseur: VWR International
Spécifications
- Dimensions:170 x 50 x 15 mm (6.7 x 1.9 x 0.59")
- Description:VWR® Cuvette Holder
- Cat. no.:89000-626
Spécifications
A propos de cet article
This polyethylene rack accommodates twelve 10 mm cuvettes, and also holds twelve cuvettes horizontally for storage and protection.
- Polyethylene construction
- Stackable for easy storage
- Space-saving design
Racks are stackable when used for storage.
Customers Who Bought This Also Bought