Magid® ROC® Polyurethane Fingertip Coated Gloves, Magid Safety
# de catalogue 69100-287
Fournisseur: Magid Glove
Spécifications
- Ambidextrous/Hand-specific:Hand-specific
- Glove Color:White
- Glove size:8
- Length:22.8 cm (9")
- Powdered/Powder-free:Powder-free
- Disposable / reusable:Reusable
- Latex/Latex-free:Latex-free
- Sterility:Non Sterile
- Gauge:15
- Cat. no.:69100-287
- Supplier No.:PU588
Spécifications
A propos de cet article
Flexible and form-fitting work gloves with polyurethane coated fingertips. The coating on the fingertips only allows the rest of the hand to breathe, as well as increasing flexibility in jobs in which the fingertip is the only area of wear.
- Scratch-resistant polyurethane coating on fingertips
- Comfortable knit wrist cuff
- Designed for lint-free applications
Gloves feature a seamless machine knit, 100% nylon shell with a knit wrist cuff. The polyurethane coating provides excellent grip, abrasion resistance, and dexterity for precise and delicate operations. This useful glove is perfect for electronics, cleanroom, and paint departments where lint is problematic.
Customers Who Bought This Also Bought