Kapak SilverPAK Pouches, Ampac Flexibles, A&D Weighing
Fournisseur: Ampac Flexibles
Certificats
A propos de cet article
Ampac's SilverPAK is a 2ply structure consisting of metalized polyester and polyethylene (METPET/PE).
These metalized pouches are ideal for powders and other granular materials.
Spécifications
- Catalog No:
- 11213-921
- 11213-954
- Material:
- Silver METPET
- Silver METPET