VWR® Cuff Tape
Fournisseur: VWR International
Certificats
A propos de cet article
This cuff tape is a conformable polyethylene tape used to ensure an effective seal between the glove and garment in cleanroom gowning.
- Low-outgassing polyethylene fabrication
- Perforated for easier tearing by hand
- Wound on a plastic core
Perforations at 3” intervals allow the user to bind and tear at the optimum length to seal the glove. The low-outgassing polyethylene film with acrylic adhesive removes cleanly leaving no adhesive residue and will not tear gloves.
Customers Who Bought This Also Bought