SP Bel-Art Single Scale Graduated Cylinders, Bel-Art Products, a part of SP
Supplier: Bel-Art Products, a Part of SP
Certificates
About this item
These graduated cylinders have a single metric scale with raised numerals for easy reading.
- Flared top rims and large spouts facilitate pouring
- Octagonal bases prevent tipping and rolling
- Non-wetting polypropylene surface
- Chemically resistant and autoclavable at 121 ºC (250 ºF)
The “non-wetting” polypropylene surface eliminates concave meniscus and makes reading easier and more accurate.
Specifications
- Catalog No:
- 70670-010
- 30617-654
- 70670-006
- 30617-652
- 70670-016
- 47750-002
- 30617-658
- 30617-656
- Form:
- Tall
- Tall
- Tall
- Tall
- Tall
- Tall
- Tall
- Tall