Pressure Reactor, Two-Piece With Bottom Outlet, Ace Glass Incorporated
Fournisseur: Ace Glass
Certificats
A propos de cet article
Two-Piece Pressure Reactor System for use at pressures up to 35 PSI at 100 Degrees C
Has threaded bottom outlet with ball joint bottom oultlet connection.
Ordering information: Complete Systems come Supplied With Valves and Ball Joint Bottom Outlet Glass Body, Schott Flange Head With #15 Center Neck,(2) #15 and (1) #7 Side Necks, Addition Funnel, Condenser, Manifold With Gauge and Rupture Disc, Stirring and Temperature Control Assemblies.
Customers Who Bought This Also Bought