SP Bel-Art No-Wire™ Cuvette Rack, Bel-Art Products, a part of SP
# de catalogue 10600-036
Fournisseur: Bel-Art Products, a Part of SP
Spécifications
- Dimensions:12.8x10.5x4.3 cm (5.1x4.1x1.7")
- Description:PP rack
- Cat. no.:10600-036
- No. of Places:42
- Supplier No.:185200000
Spécifications
A propos de cet article
All-polypropylene rack provides gentle no-scratch support for fragile and expensive spectrophotometer cuvettes.
- Polypropylene construction
- Will not scratch glass cuvettes
- Autoclavable
Forty-two 10 mm light path cuvettes fit in this 6 x 7 row layout. Steam autoclavable at 121ºC (250ºF).
Customers Who Bought This Also Bought