SP Bel-Art 'C'-Shape Open Lead Ring Flask Weights with Vikem® Vinyl Coating, Bel-Art Products, a part of SP
Fournisseur: Bel-Art Products, a Part of SP
Certificats
A propos de cet article
These rings fit neatly around the neck of filtering flasks with side-arm tubing connections without disconnecting the tubing.
- Green vinyl coating
- Flexible
- Available in four diameter/weight combinations
The open ring, made of lead and coated with green Vikem® vinyl to protect glassware, is flexible enough to be pulled apart to accommodate a wider vessel.
Customers Who Bought This Also Bought