SP Bel-Art Heavy Duty Rectangular Tanks with Top Flanges, Bel-Art Products, a part of SP
Fournisseur: Bel-Art Products, a Part of SP
Certificats
A propos de cet article
Seamless, rotational-molded polyethylene tanks are lightweight and easy to handle.
- Polyethylene construction
- Seamless rotational mold
- Lightweight
- Easy to handle
Available with or without a faucet, these tanks are resistant to most acids and all alkalis. All sizes are made with wide top flanges, include covers.
Ordering information: Can be supplied with spigots, fittings, dams, partitions etc. Contact your Avantor representative for quotations on special sizes and fabrication in polypropylene and PVC.
Customers Who Bought This Also Bought