Glass Check Valve with Ground Seat, Ace Glass Incorporated
# de catalogue 89064-042
Fournisseur: Ace Glass
Product available on GSA Advantage®
Small Business Enterprise
Spécifications
- Description:With Ground Seat
- Cat. no.:89064-042
- Supplier No.:8774-09
Spécifications
A propos de cet article
All-glass check valve with ground seat and precision ground solid ball
Suitable with mercury in the vertical position, more sensitive to other fluids in the horizontal position. Connections are 8 mm O.D.
Customers Who Bought This Also Bought