Single Tube Vacuum Manifold, #15 Ace-Thred Joint Connections and PTFE Stopcocks, Ace Glass Incorporated
Fournisseur: Ace Glass
Certificats
A propos de cet article
Glass manifold with no.15 Ace-Thred at either end to allow switching vacuum connection to left or right side
Supplied with three or five 4 mm straight bore PTFE stopcock sampling ports. Overall length of glass manifold is 230 mm.
Delivery: Complete item consists of a glass manifold, 89052-848 end plug with FETFE o-ring, and 89052-948 and 89052-954 Ace-Safe connector with bushing for 1/4" or 1/2" tubing connection.
Customers Who Bought This Also Bought