One-Piece Unjacketed Pressure Reactor, Ace Glass
Fournisseur: Ace Glass
Certificats
A propos de cet article
Rugged one-piece heavy wall unjacketed pressure reactor, for use at pressure of up to 45 psig @ 100°C
Designed to perform low to moderate positive pressure reactions, synthesis and catalysis. Reactor capacities range from 500 mL to 5000 mL and use Ace-Threds to achieve a leak-tight system. One-piece design features a 25 mm center opening and has a higher allowable pressure rating than our two-piece design.
Ordering information: System option comes complete with (2) #15 PTFE Plugs, Condenser, Flask, Head, Manifold, Thermowell, Clamp, Funnel, (2) #15 PTFE Bushing, bearing w/ Ace-threads, Stir Shaft, Flexible Shaft, PTFE Stirrer Blade, Krytox High Vacuum Grease, Swivel Coupling, Polypropylene Tubing, Motor Controller, Mounting Rod, PTFE Sealing Tape.
Customers Who Bought This Also Bought