SP Bel-Art Vacuum Aspirator Pump, Bel-Art Products, a part of SP
# de catalogue 13197-066
Fournisseur: Bel-Art Products, a Part of SP
Spécifications
- Description:Vacuum Aspirator Pump
- Cat. no.:13197-066
- Supplier No.:329470000
Spécifications
A propos de cet article
High-density polyethylene aspirator pump operates on water pressure above 11psi (7584 N/m2).
- HDPE construction
- Maximum vacuum of 69.6 cm Hg
- Corrosion-resistant check valve
The integral check valve resists corrosive filtrates and fumes. A quick-disconnect hose fitting accepts 6.4 to 9.5 mm (¼ to ⅜”) I.D. tubing. The male thread faucet connection is 9.5 mm (⅜”) I.D. Maximum attainable vacuum is 69.6 cm Hg (27.4”) with a flow of 6 liters (1.5 gallons) per minute.
Customers Who Bought This Also Bought