VACUSIP Bench-Top Aspiration System, INTEGRA Biosciences
Fournisseur: INTEGRA Biosciences
Certificats
A propos de cet article
VACUSIP provides an out-of-the-box solution for the wide array of liquid waste disposal tasks carried out at the laboratory bench
- Reliable low noise pump, enabling it to operate independently from an external vacuum source
- Rechargeable battery version provides even more flexibility
- Unique hand operator allows a fine regulation of the aspiration flow and accepts a variety of adapters
The 0.5 L glass bottle is quickly exchanged and all components coming in contact with liquids are autoclavable.
Battery capacity (when fully charged): 5 days when aspirating 1 h per day (model 89498-930 with battery only).
Certifications: CE certified.
Ordering information: Aspiration system with borosilicate bottle, silicone hand operator (incl. 1- and 8-channel steel tips and 1-channel adapter for standard tips/GRIPTIPS), 1.2 m silicone tubing, filter and mains adapter.
Spécifications
- Catalog No:
- 89498-930
- 89498-932
- Bottle capacity:
- Borosilicate with GL45 lid, 500 mL
- Borosilicate with GL45 lid, 500 mL
- Dimensions:
- 125H x 120W x 162D mm (4.92 x 4.72 x 6.38"); 345 mm (13.6") height with bottle and aspiration device
- 125H x 120W x 162D mm (4.92 x 4.72 x 6.38"); 345 mm (13.6") height with bottle and aspiration device
- Filters:
- 0.45 μm hydrophobic filter
- 0.45 μm hydrophobic filter
- Vacuum range:
- -250 mbar ±20 %
- -250 mbar ±20 %
- Weight:
- 500 g (1.1 lb.) without bottle
- 500 g (1.1 lb.) without bottle