Glass or PTFE Baffles, Ace Glass Incorporated
Supplier: Ace Glass
Certificates
About this item
Use Ace-Thred adapters to mate glass or PTFE (stainless steel core) baffles to any standard taper joint
Baffles can be adjusted vertically or rotated to increase agitation.
Customers Who Bought This Also Bought