Stoppers, PTFE, Ace Glass Incorporated
Fournisseur: Ace Glass
Certificats
A propos de cet article
Virgin TFE/PTFE stoppers with built-in black Delrin stopper extractor ring
Eliminates the hazard of removing a frozen stopper from glass vessel. You need only turn red nut clockwise to exert a gentle force against the vessel’s lip and the non-stick stopper is extracted.
Customers Who Bought This Also Bought