Double Tube Vacuum Manifold, O-Ring Joint Connections and Easy Action Valves, Ace Glass Incorporated
Fournisseur: Ace Glass
Certificats
A propos de cet article
The no.15 o-ring joint connections at each end
Three or four ports with easy action 0 - 10 mm threaded stopcocks and standard taper joints or no.15 o-ring joint connections off each port. The distance between ports is 100 mm, which will accept 250 mL flasks or smaller. Requires all stainless steel, pinch type screw locking clamp 89060-256 (for no.15 o-ring joint).
Customers Who Bought This Also Bought