Automatic Pressure Release Valve, Adjustable, Ace Glass Incorporated
Fournisseur: Ace Glass
Certificats
A propos de cet article
Automatic, adjustable pressure release valve for safer operation of pilot plant or other assemblies where the pressure is of great concern
The tension on the stainless steel spring determines the amount of pressure needed to force the lower piece open. With the spring supplied, the range is 15–50 psi. Available with or without warning whistle. Plunger made of PTFE with FETFE o-rings.
Customers Who Bought This Also Bought