Ultra-Precision NMR Sample Tubes, 300Mhz, Ace Glass Incorporated
Fournisseur: Ace Glass
Certificats
A propos de cet article
Ultra precise and accurate, borosilicate glass NMR tubes are available in 17.8 cm (7") and 20.3 cm (8") lengths with a 5 mm Ext.Ø.
Customers Who Bought This Also Bought