Glass Check Valve with Ground Seat for Liquid Seal, Ace Glass Incorporated
Fournisseur: Ace Glass
Certificats
A propos de cet article
An all-glass check valve with ground seat for liquid seal
Available in style A — for vertical use, and style B — for right angle connection. Connections are 8 mm O.D.
Customers Who Bought This Also Bought