Double Tube Vacuum Manifold, O-Ring Joint Connections and Glass Stopcocks, Ace Glass Incorporated
Fournisseur: Ace Glass
Certificats
A propos de cet article
The no.15 o-ring joint connections at each end
Three or four ports with 4 mm bore glass double oblique stopcocks and standard taper joints or no.15 o-ring joint connections off each port. The distance between ports is 210 mm, which will accept 3 L flasks or smaller. Requires all-stainless-steel pinchtype screw locking clamp 89060-256.
Customers Who Bought This Also Bought