Pressure Reactor Moisture Test Receiver, Ace Glass Incorporated
Fournisseur: Ace Glass
Certificats
A propos de cet article
Used to determine heavier-than-water, co-distilling solvent component content of reaction mixture
For use with ACE pressure reactors with #15 Ace-Thred. Side arm is connected to #15 Ace-Thred on pressure reactor head with a bushing and size –110 FETFE o-ring, 89060-984 . The 89054-000 pressure condenser, with #15 top plug, is attached to #15 thread at top of receiver with bushing and o-ring. Receiver capacity is 60 mL, graduated in 1 mL subdivisions. Bottom outlet stopcock is a 0-3 mm with FETFE o-rings. Pressure rating, 45 psig @100°C. Complete consists of receiver and bushing with o-ring. Condenser and #15 Plug NOT supplied — order separately.
Customers Who Bought This Also Bought