VWR® Flexible Safety Caps
Fournisseur: VWR International
Certificats
A propos de cet article
Safety caps provide a safe and economical way to cap and recap blood collection tubes and disposable glass culture tubes, regardless of manufacturer.
- Caps are color-coded for easy identification
- Liquid-tight seals
- Safe and economical
VWR® Flexible Safety Caps are available for a wide range of biomedical and industrial applications and fit six different sized tubes. VWR® Flexible Safety Caps come in a variety of colors for coding samples. They feature liquid-tight seals to ensure sample integrity and prevent spillage of serum samples. Made in the USA.
Customers Who Bought This Also Bought