Graduated Transfer Pipettes, Globe Scientific
Fournisseur: Globe Scientific
Certificats
A propos de cet article
Produced from a custom grade low-density polyethylene (LDPE), these unbreakable one-piece pipettes are used to draw and dispense liquids in controlled, reproducible drop sizes.
- Non-toxic and chemically inert to biological fluids and most acids
- Easy to grip with gloves
- Temperature Range: –50 to 90 °C
- Can be gas (EtO) sterilized
Designed for measurements, these graduated transfer pipettes feature molded-in graduation marks that will not fade or wash off.
Customers Who Bought This Also Bought