Large Scale Evaporator Flasks, Ace Glass Incorporated
Fournisseur: Ace Glass
Certificats
A propos de cet article
These large flasks are from blanks selected for balance and quality
Necks are carefully welded to prevent “rotational whip.” Flasks can be plastic coated* upon request.
Customers Who Bought This Also Bought